missing translation for 'onlineSavingsMsg'
Learn More

Magnesium ribbon, approximate dimensions, 3.0mm (0.12in) wide, 0.15mm (0.0059in) thick, Thermo Scientific Chemicals

Product Code. 15404755
Click to view available options
Quantity:
25 g
5 x 25 g
Unit Size:
25g
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 15404755

Brand: Thermo Scientific Alfa Aesar 012811.51

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Form: approx. 0.78g/m; Highly flammable

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7439-95-4
Molecular Formula Mg
Molecular Weight (g/mol) 24.30
MDL Number MFCD00085308
InChI Key JLVVSXFLKOJNIY-UHFFFAOYSA-N
Synonym magnesio, sheet, powdered, magnesio italian, turnings, rieke's active, ribbon, unii-i38zp9992a, hsdb 654, nitrate, acs
PubChem CID 5462224
ChEBI CHEBI:25107
IUPAC Name magnesium
SMILES [Mg]

Specifications

Quantity 5 x 25 g
Odor Odorless
Weight ≈0.78g/m
UN Number UN1869
Chemical Name or Material Magnesium ribbon, Approximate dimensions: 3.0mm (0.12 in.) wide, 0.15mm (0.0059 in.) thick

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.