missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Magnesium rod, 6.35mm (0.25in) dia x 25.4mm (1.0in) length, 99.95% (metals basis), Thermo Scientific Chemicals
Click to view available options
Quantity:
1 Pc.
25 Pc.
5 Pc.
Unit Size:
Each
Pack of 25
Pack of 5
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Quantity | 5 Pc. |
| Odor | Odorless |
| Weight | ≈1.4g/piece |
| Percent Purity | 99.95% |
| Assay | (metals basis) |
| Chemical Name or Material | Magnesium rod, 6.35mm (0.25 in.) dia. x 25.4mm (1.0 in.) length |
Safety and Handling
Product Identifier
- Magnesium rod
- Danger
- Flammable solid Category 1
- Self-heating substance Category 2
- Substance which\, in contact with water\, emits flammable gases Category 2
- H228-Flammable solid.
- H252-Self-heating in large quantities; may catch fire.
- H261-In contact with water releases flammable gases.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P223-Do not allow contact with water.
- MIXTURE LIST-Contains : Magnesium
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction