missing translation for 'onlineSavingsMsg'
Learn More
Magnesium sulfate, 97%, pure, anhydrous, Thermo Scientific Chemicals Product Code.: 10003812

Magnesium sulfate, 97%, pure, anhydrous, Thermo Scientific Chemicals

Product Code. 10003812
2.5 kg, Plastic Bottle
Click to view available options
Quantity:
5 g
500 g
2.5 kg
25 kg
Packaging:
Fibre drum
Glass Bottle
Plastic Bottle
This item is not returnable. View return policy

Product Code. 10003812

Brand: Thermo Scientific Acros 413480025

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Magnesium sulfate, 97%, MgO4S, CAS Number-7487-88-9, sulfuric acid magnesium salt 1:1, sal de sedlitz, magnesium sulfate, tomix ot, bitter salt, magnesium sulfate 1:1, sal angalis, magnesium sulphate, mgso4, magnesium sulfate anhydrous, 500g, CHEBI:32599, Undesignated, >700 deg.C

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Adsorbent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7487-88-9
Molecular Formula MgO4S
Molecular Weight (g/mol) 120.36
MDL Number MFCD00011110
InChI Key CSNNHWWHGAXBCP-UHFFFAOYSA-L
Synonym magnesium sulfate, magnesium sulphate, magnesium sulfate anhydrous, sulfuric acid magnesium salt 1:1, bitter salt, sal angalis, magnesium sulfate 1:1, mgso4, sal de sedlitz, tomix ot
PubChem CID 24083
ChEBI CHEBI:32599
IUPAC Name magnesium;sulfate
SMILES [Mg++].[O-]S([O-])(=O)=O

Specifications

Melting Point 1124.0°C
Physical Form Granular Powder and/or Specks
Quantity 2.5 kg
Assay Percent Range 96% min. (Complexometry)
Linear Formula MgSO4
Fieser 01,634; 04,425; 05,421
Merck Index 15, 5755
Solubility Information Solubility in water: 26g/100mL (0°C).
Formula Weight 120.37
Percent Purity 97%
Grade Pure
Packaging Plastic Bottle
Decomposition Information >700°C
Chemical Name or Material Magnesium sulfate, Pure
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0423143 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number413480050,413480250,413485000,413480025
Lot NumberA0468734 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number413480050,413480250,413485000,413480025
Lot NumberA0465493 Certificate TypeCertificate of Analysis Date19/08/2024 Catalog Number413480050,413480250,413485000,413480025
Lot NumberB0155501B Certificate TypeCertificate of Analysis Date11/06/2024 Catalog Number413480050,413480250,413485000,413480025
Lot NumberB0155501 Certificate TypeCertificate of Analysis Date11/06/2024 Catalog Number413480050,413480250,413485000,413480025

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Magnesium sulfate, 97%, pure, anhydrous, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.