missing translation for 'onlineSavingsMsg'
Learn More

Magnesium sulfate, 99%, extra pure, dried, contains 3 to 4 moles of water, Thermo Scientific Chemicals

Product Code. 10256350
Click to view available options
Quantity:
1 kg
2.5 kg
10 kg
Packaging:
Plastic Bottle
Plastic drum

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Adsorbent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22189-08-8
Molecular Formula MgO4S
Molecular Weight (g/mol) 120.36
MDL Number MFCD00149787
InChI Key CSNNHWWHGAXBCP-UHFFFAOYSA-L
Synonym magnesium sulfate, magnesium sulphate, magnesium sulfate anhydrous, sulfuric acid magnesium salt 1:1, bitter salt, sal angalis, magnesium sulfate 1:1, mgso4, sal de sedlitz, tomix ot
PubChem CID 24083
ChEBI CHEBI:32599
SMILES [Mg++].[O-]S([O-])(=O)=O

Specifications

Melting Point 1124.0°C
Color White
Physical Form Crystalline Powder
Quantity 1 kg
Assay Percent Range 98.5% min. (On dried substance) (Complexometry)
Linear Formula MgSO4·xH2O
Solubility Information Solubility in water: 360g/L (20°C). Other solubilities: soluble in glycerol, sparingly soluble in alcohol
IUPAC Name magnesium(2+) sulfate
Formula Weight 120.36
Percent Purity 99%
Grade Extra Pure
Loss on Drying 26% to 32% (400 to 500 °C)
Packaging Plastic Bottle
Specific Gravity 2.66
Density 2.6600g/mL
Chemical Name or Material Magnesium sulfate, Dried, contains 3 to 4 Moles of Water
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'