missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Magnesium Sulfate, Extra Pure, SLR, Dried, Fisher Chemical™
34.00 € - 159.00 €
Chemical Identifiers
CAS | 22189-08-8 |
---|---|
Molecular Formula | MgO4S |
Molecular Weight (g/mol) | 120.36 |
MDL Number | MFCD00149787 |
InChI Key | CSNNHWWHGAXBCP-UHFFFAOYSA-L |
Synonym | magnesium sulfate, magnesium sulphate, magnesium sulfate anhydrous, sulfuric acid magnesium salt 1:1, bitter salt, sal angalis, magnesium sulfate 1:1, mgso4, sal de sedlitz, tomix ot |
PubChem CID | 24083 |
ChEBI | CHEBI:32599 |
SMILES | [Mg++].[O-]S([O-])(=O)=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10316240
|
Fisher Chemical
M/1100/53 |
500 g | Plastic powder jar |
34.00 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10776101
|
Fisher Chemical
M/1100/60 |
1 kg | Plastic powder jar |
54.00 €
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10224680
|
Fisher Chemical
M/1100/61 |
2 kg | Plastic powder jar |
88.35 €
2kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10181860
|
Fisher Chemical
M/1100/68 |
10 kg | Plastic keg |
159.00 €
10kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Magnesium Sulfate, Extra Pure, SLR, Dried
Chemical Identifiers
22189-08-8 | |
120.36 | |
CSNNHWWHGAXBCP-UHFFFAOYSA-L | |
24083 | |
[Mg++].[O-]S([O-])(=O)=O |
MgO4S | |
MFCD00149787 | |
magnesium sulfate, magnesium sulphate, magnesium sulfate anhydrous, sulfuric acid magnesium salt 1:1, bitter salt, sal angalis, magnesium sulfate 1:1, mgso4, sal de sedlitz, tomix ot | |
CHEBI:32599 |
Specifications
1124°C | |
White | |
MgO4S | |
magnesium sulfate, magnesium sulphate, magnesium sulfate anhydrous, sulfuric acid magnesium salt 1:1, bitter salt, sal angalis, magnesium sulfate 1:1, mgso4, sal de sedlitz, tomix ot | |
[Mg++].[O-]S([O-])(=O)=O | |
120.36 | |
CHEBI:32599 | |
<0.1mmHg at 20°C |
22189-08-8 | |
Powder/Solid | |
MFCD00149787 | |
CSNNHWWHGAXBCP-UHFFFAOYSA-L | |
magnesium;sulfate | |
24083 | |
120.36g/mol |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title