missing translation for 'onlineSavingsMsg'
Learn More

Magnesium Sulfate, Reagent grade, Honeywell

Código de producto. 15661820
missing translation for 'orderingAttributeHoverText'
Quantity:
500 g
2 kg
2.5 kg
5 kg
50 kg

Reagent Grade, anhydrous,≥97%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Identificadores de productos químicos

CAS 7487-88-9
Molecular Formula MgO4S
Molecular Weight (g/mol) 120.36
MDL Number MFCD00011110
InChI Key CSNNHWWHGAXBCP-UHFFFAOYSA-L
Synonym magnesium sulfate, magnesium sulphate, magnesium sulfate anhydrous, sulfuric acid magnesium salt 1:1, bitter salt, sal angalis, magnesium sulfate 1:1, mgso4, sal de sedlitz, tomix ot
PubChem CID 24083
ChEBI CHEBI:32599
SMILES [Mg++].[O-]S([O-])(=O)=O

Especificaciones

Physical Form Anhydrous
Quantity 2.5 kg
UN Number NONH for all modes of transport
IUPAC Name magnesium;sulfate
Formula Weight 120.37g/mol
Percent Purity ≥97%
Grade Reagent
Vapor Pressure 0.1mmHg (20°C)
Density Vapor Density: <0.01 (vs air)
Chemical Name or Material Magnesium Sulfate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'