missing translation for 'onlineSavingsMsg'
Learn More

Magnesium Sulfate Solution, 1.0 M, Honeywell™

Product Code. 15626080
Click to view available options
Quantity:
1 L

Volumetric, 1.0 M MgSO4

Honeywell´s high-quality titration products cover the complete titration workflow and include:

  • Solutions for volumetric titration (acids, bases, or salts)
  • Buffers available as concentrates or ready-to-use

 

  • Reagents for complexometry:
    • Aminopolycarboxylic acids (EDTA/NTA analogs)
    • Masking agents
  • Indicators for volumetric and complexometric titrations

 


 

TRUSTED_SUSTAINABILITY

Specifications

CAS 7487-88-9
Quantity 1 L
Molecular Formula MgO4S
MDL Number MFCD00011110
UN Number NONH for all modes of transport
Synonym magnesium sulfate, magnesium sulphate, magnesium sulfate anhydrous, sulfuric acid magnesium salt 1:1, bitter salt, sal angalis, magnesium sulfate 1:1, mgso4, sal de sedlitz, tomix ot
InChI Key CSNNHWWHGAXBCP-UHFFFAOYSA-L
SMILES [Mg++].[O-]S([O-])(=O)=O
IUPAC Name magnesium;sulfate
Molecular Weight (g/mol) 120.36
PubChem CID 24083
ChEBI CHEBI:32599
Formula Weight 120.37g/mol
Chemical Name or Material Magnesium sulfate solution
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'