missing translation for 'onlineSavingsMsg'
Learn More

Magnesium turnings, 1cm (0.4in) & down, Puratronic™, 99.98% (metals basis), Thermo Scientific Chemicals

Product Code. 11340819
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7439-95-4
Molecular Formula Mg
Molecular Weight (g/mol) 24.30
MDL Number MFCD00085308
InChI Key JLVVSXFLKOJNIY-UHFFFAOYSA-N
Synonym magnesio, sheet, powdered, magnesio italian, turnings, rieke's active, ribbon, unii-i38zp9992a, hsdb 654, nitrate, acs
PubChem CID 5462224
ChEBI CHEBI:25107
IUPAC Name magnesium
SMILES [Mg]

Specifications

UN Number UN1869
Quantity 5 g
Percent Purity 99.98%
Grade Puratronic™
Odor Odorless
Particle Size 1cm (0.4 in.) & down
Assay (metals basis)
Chemical Name or Material Magnesium turnings

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'