Learn More
3M™ Magnifying Lens
3M™ Magnifying Lens is used for magnifying object and can be used with all Speedglas welding helmets.
Brand:  3M™ 7000028023
21.36 EUR valid until 2025-01-31
Use promo code "23894" to get your promotional price.
 View more versions of this product
Product Code. 15869634
  • 28.00 € / Each
Estimated Shipment: 21-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • On the 3M™ Speedglas™ Welding Helmet Series 9100 there is a magnifying lens holder built into the welding filter, making lens installation easier and more secure
  • For the new 3M™ Speedglas™ Welding Helmet Series 100 there is a magnifying lens holder built into the welding filter, making lens installation easier and more secure. For the older 100 series filters you will need a holder for the magnifier
  • For the 3M™ Speedglas™ Welding Helmet SL you will need to add the holder for magnifier as a separate spare part (171031)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Speedglas
2.0X
Magnifying Lens