Learn More
Interscience™ Magnifying Lens
Magnifying lens for close inspection and counting of colonies.
Brand:  Interscience™ 435001
Product Code. 12628685
  • 315.00 € / Each
Explore more special offers
Description
Description
  • Includes magnifying lens and flexible arm for Scan 50, Scan 50 Pro and Scan 100
Specifications
Specifications
Transparent
Scan 50, 50 Pro, Scan 100
Glass
Magnifying Colonies
2x
Lens, Magnifying