missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ MagPette™ Pipette Holder
GREENER_CHOICE

Secures to most metal surfaces with four powerful magnets. Fisherbrand™ MagPette™ holds two pipetters and fits most pipetter brands.

Brand:  Fisherbrand™ HS23503

14.36 EUR valid until 2024-12-31
Use promo code "21745" to get your promotional price.


Product Code. 11932581

  • 23.10 € / Each
In Stock
to see stock.
Edge
Explore more special offers
Add to basket

Description

Description

  • Constructed of robust ABS plastic
  • Available in bright translucent colors.
  • Dimensions: (W x D x H) 131 x 112 x 76 mm
Specifications

Specifications

Pipette Rack
Pipetters
2
131 mm
3 in.
112 mm
Purple
ABS
Magnet
76 mm
4.4 in.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.