missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3M™ Maintenance Free Half Mask
This Maintenance Free Half Mask is a simple and ready-to-use respirator with an integrated cartridge and filters.
39.60 € - 49.75 €
Specifications
Certifications/Compliance | EN 405:2001, A1:2009 |
---|---|
Certifications (EN) | EN 405:2001+A1:2009 |
Connection Type | Permanent |
Color | Blue |
Design Type | Half Mask |
Product Code | Brand | Model | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Model | Price | Quantity & Availability | ||||||
16486350
|
3M™
7100113098 |
4251+ |
39.60 €
Each |
In Stock
Log in to see stock. ![]() |
||||||
16496350
|
3M™
7100113101 |
4255+ |
49.75 €
Each |
In Stock
Log in to see stock. ![]() |
||||||
16406360
|
3M™
7100113102 |
4277+ |
48.60 €
Each |
In Stock
Log in to see stock. ![]() |
||||||
Description
- Effective and comfortable protection against organic vapor, inorganic vapor, acid gases, ammonia, and hazardous particulates.
- A newly enhanced valve assembly helps to reduce exhalation breathing resistance for improved breathing comfort.
- Lightweight, well-balanced, low profile design.
- Integrated cartridge and filters mean it's ready to use and maintenance free.
- Soft, close-fitting face seal made with textured, non-allergenic material.
- Includes a resealable bag for hygienic storage and transportation.
Specifications
EN 405:2001, A1:2009 | |
Permanent | |
Half Mask | |
Bonded Carbon | |
Reusable Respirator | |
Yes | |
Polyphenylene Ether |
EN 405:2001+A1:2009 | |
Blue | |
Reusable | |
Thermoplastic Elastomer | |
One Size | |
TPE | |
Polyisoprene |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
3M™ Maintenance Free Half Mask