Learn More
Rogo Sampaic™ Malassez Counting Cells
For determining the number of particles per volume unit of a liquid. Rogo Sampaic™ Malassez Counting Cells have a simple or double grid with a depth of 0.2mm.
Brand:  Rogo Sampaic™ ZMB021
 View more versions of this product
Product Code. 11720845
  • 157.00 € / Each
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
Description
Description
  • For blood samples
  • Provides for accurate manual measurements
Specifications
Specifications
Malassez Cell
1Each
0.2mm
Double
Blood
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Rogo Sampaic™ Malassez Counting Cells > Grid Type: Double