missing translation for 'onlineSavingsMsg'
Learn More

Brand™ Counting Chambers, Malassez pattern, BLAUBRAND™, Double ruling

BLAUBRAND™ quality. Outstanding measurement precision. 100% certified quality.

Brand:  Brand™ 719005

59.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.


Product Code. 10523281

  • 62.92 € / Each
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Manufactured according to DIN 12 847, including 2 haemacytometer cover glasses.
  • The ruling is rectangular, covering an area of 5 mm2.
  • The large rectangles measure 0.25 x 0.20 = 0.05 mm2.
  • They are each subdivided into 20 mini squares with an area of 0.0025 mm2.
  • This chamber is commonly used for counting cells in the cerebro-spinal fluid, or for counting nematodes.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Counting Device
1Each
0.2mm
Counting cells in the cerebro-spinal fluid, or nematodes.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Brand™ Counting Chambers, Malassez pattern, BLAUBRAND™, Double ruling > Without Spring Clips

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.