Learn More
Value Plastics Male Luer Integral Lock Ring to NPT(M) Thread Adapters
Shop All Cole-Parmer Products
Click to view available options
Thread Size:
1/4-18 NPT(M)
1/8-27 NPT(M)
Material (Seal):
Nylon
Polypropylene
Unit Size:
Pack of 25
Description
Easily transition from a luer to thread fitting. Luer fittings allow for a simple yet secure twist-lock connection. NPT thread fittings are designed to create a tight seal under pressure
Specifications
Specifications
| Certifications/Compliance | ADCF;FDA;Non-DEHP;Non-Phthalate;RoHS;USP |
| Thread Size | 1/4-18 NPT(M) |
| Material (Seal) | Polypropylene |
| Product Type | Male Luer Integral Lock Ring Hex |