Learn More
Hamilton™ Male Luer Lock M6 Threads
Hamilton offers a diverse line of inert fittings fittings and connectors made from CTFE. The fittings are designed for use with industry standard luer and luer lock products like our syringes as well as threaded 1/4-28 and M6 flat bottom ports.
Brand:  Hamilton™ 32876
Product Code. 11533244
  • 60.00 € / Each
Estimated Shipment: 20-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Specification Sheet
Connection 1Male Luer Lock
Connection 2Male M6
Sales Quantity1 each
I.D.1.5 mm
Specifications
Specifications
Male Luer Lock M6 Threads