Learn More
Idex Male-Threaded Nuts
Combine with Super Flangeless™ ferrule for a high pressure holding flat-bottom fitting assembly
POA - 81.00 €
Product Code | Brand | For Use With (Equipment) | Length (English) | Material | Width (English) | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | Length (English) | Material | Width (English) | Price | Quantity & Availability | ||
13070950
|
Idex
U-400X |
1/16 in. O.D. Tubing | 0.4 in. | Stainless Steel | 0.25 in. | N/A | |||
12977589
|
Idex
02013-73 |
1/32 in. or 1/16 in. OD Tubing | 0.87 in. | PEEK | 0.365 in. |
81.00 €
Pack of 10 |
In Stock
Log in to see stock. |
||
12917629
|
Idex
02021-80 |
1/8 in. O.D. Tubing | 0.87 in. | PEEK | 0.365 in. |
8.85 €
Each |
In Stock
Log in to see stock. |
||
Description
Combine with Super Flangeless™ ferrule for a high pressure holding flat-bottom fitting assembly
- Eliminates tubing twist that can loosen connections
- Resistant to loosening through vibration