missing translation for 'onlineSavingsMsg'
Learn More

Maleate, 0.2M buffer soln., pH 6.0, Thermo Scientific™

46.00 € - 61.00 €

Chemical Identifiers

CAS 3105-55-3
Molecular Formula C4H2NaO4
Molecular Weight (g/mol) 137.05
MDL Number MFCD00070246
InChI Key VRVKOZSIJXBAJG-ODZAUARKSA-L
PubChem CID 91658932
SMILES [Na+].[O-]C(=O)\C=C/C([O-])=O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15494489
View Documents Promotion Details
Thermo Scientific Alfa Aesar
J62035.AK
250 mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15404499
View Documents Promotion Details
Thermo Scientific Alfa Aesar
J62035.AP
500 mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

3105-55-3
137.05
VRVKOZSIJXBAJG-ODZAUARKSA-L
[Na+].[O-]C(=O)\C=C/C([O-])=O
C4H2NaO4
MFCD00070246
91658932

Specifications

3105-55-3
0.2 M
Liquid
MFCD00070246
[Na+].[O-]C(=O)\C=C/C([O-])=O
137.05
Maleate buffer soln.
Colorless
6.0
C4H2NaO4
VRVKOZSIJXBAJG-ODZAUARKSA-L
sodium (2Z)-but-2-enedioate
91658932
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 221-461-6

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications