missing translation for 'onlineSavingsMsg'
Learn More

Maleate, 0.2M buffer soln., pH 6.5, Thermo Scientific™

50.05 € - 72.70 €

Chemical Identifiers

CAS 676-46-0
Molecular Formula C4H4Na2O5
Molecular Weight (g/mol) 178.051
MDL Number MFCD00012466
InChI Key WPUMTJGUQUYPIV-UHFFFAOYSA-L
PubChem CID 8736
IUPAC Name disodium;2-hydroxybutanedioate
SMILES C(C(C(=O)[O-])O)C(=O)[O-].[Na+].[Na+]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15443789
View Documents
Thermo Scientific Alfa Aesar
J61156.AK
250 mL
50.05 €
250mL
Please to purchase this item. Need a web account? Register with us today!
15453789
View Documents
Thermo Scientific Alfa Aesar
J61156.AP
500 mL
72.70 €
500mL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

676-46-0
178.051
WPUMTJGUQUYPIV-UHFFFAOYSA-L
disodium;2-hydroxybutanedioate
C4H4Na2O5
MFCD00012466
8736
C(C(C(=O)[O-])O)C(=O)[O-].[Na+].[Na+]

Specifications

676-46-0
6.5
C4H4Na2O5
WPUMTJGUQUYPIV-UHFFFAOYSA-L
disodium;2-hydroxybutanedioate
8736
0.2 M
Liquid
MFCD00012466
C(C(C(=O)[O-])O)C(=O)[O-].[Na+].[Na+]
178.051
Maleate buffer soln.
Videos
Safety and Handling

Safety and Handling

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications