missing translation for 'onlineSavingsMsg'
Learn More

Maleic Acid, 99%, Thermo Scientific Chemicals

Product Code. 10439050
Click to view available options
Quantity:
100 g
1 kg
5 kg
25 kg
Packaging:
Fibre drum
Plastic bottle
Plastic drum
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110-16-7
Molecular Formula C4H4O4
Molecular Weight (g/mol) 116.07
MDL Number MFCD00063177
InChI Key VZCYOOQTPOCHFL-OWOJBTEDSA-N
Synonym maleic acid, cis-butenedioic acid, toxilic acid, maleinic acid, malenic acid, 2z-but-2-enedioic acid, 2-butenedioic acid z, maleate, cis-1,2-ethylenedicarboxylic acid, z-butenedioic acid
PubChem CID 444266
ChEBI CHEBI:18300
IUPAC Name (Z)-but-2-enedioic acid
SMILES OC(=O)\C=C\C(O)=O

Specifications

Melting Point 134.0°C to 138.0°C
Color White
Assay Percent Range 99%
Linear Formula HO2CCH=CHCO2H
Beilstein 02, IV, 2199
Merck Index 15, 5767
Solubility Information Solubility in water: 790g/L (25°C). Other solubilities: soluble in glacial acetic acid, soluble in alcohol and acetone, slightly soluble in ether and benzene, practically insoluble in carbon tetrachloride
Formula Weight 116.07
Percent Purity 99%
Quantity 1 kg
Flash Point 127°C
Infrared Spectrum Authentic
Packaging Plastic bottle
Physical Form Crystalline Powder
Chemical Name or Material Maleic acid
Show More Show Less
compliance-icons
Product Identifier
  • Maleic acid
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'