missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Maleic anhydride, briquettes, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
1 kg
5 kg
25 kg
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Spécification
Melting Point | 52.0°C to 55.0°C |
Boiling Point | 200.0°C |
Flash Point | 102°C |
Quantity | 25 g |
Solubility Information | Solubility aq. soln.: 400 g/L (25°C). Other solubilities: reacts with water forming maleic acid, 227 g/100 g acetone - 50 g/100 g benzene (25°C), 112 g/100 g ethylacetate (25°C)- 0,60 g/100 g ccl4, 52.5 g/100 g chloroform (25°C)-23,4 g/100 g toluen |
Viscosity | 1.6 mPa.s (60°C) |
Formula Weight | 98.06 |
Chemical Name or Material | Maleic anhydride |
RUO – Research Use Only
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu