missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cadence Science™ Malleable Stainless Steel Animal Feeding Needles
Designed to meet the special needs of pharmacologists, toxicologists, pathologists, and other biomedical researchers working with laboratory animals. Cadence Science™ Malleable Stainless Steel Animal Feeding Needles are sterile and disposable.
Brand: Cadence Science™ 9921
This item is not returnable.
View return policy
Description
- Type 304 stainless steel construction
- Malleable
- Sterile
- Disposable
- 100/box, 20 x 5/sterile pouch
Specifications
Animal Feeding Needle | |
Single-use | |
20G | |
1.5 in. |
Stainless Steel (304) | |
Animal feeding | |
Sterile | |
1.9 mm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction