Learn More
Malononitrile, 99%, Thermo Scientific Chemicals
27.03 € - 499.49 €
Chemical Identifiers
CAS | 109-77-3 |
---|---|
Molecular Formula | C3H2N2 |
MDL Number | MFCD00001883 |
InChI Key | CUONGYYJJVDODC-UHFFFAOYSA-N |
Synonym | malononitrile, dicyanomethane, cyanoacetonitrile, malonic dinitrile, methylene cyanide, dicyanmethane, malonodinitrile, malonic acid dinitrile, methylenedinitrile, methane, dicyano |
PubChem CID | 8010 |
ChEBI | CHEBI:33186 |
IUPAC Name | propanedinitrile |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11408840
|
Thermo Scientific Alfa Aesar
A15046.06 |
5 g |
27.03 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11467307
|
Thermo Scientific Alfa Aesar
A15046.22 |
100 g |
29.04 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11477307
|
Thermo Scientific Alfa Aesar
A15046.36 |
500 g |
93.57 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11487307
|
Thermo Scientific Alfa Aesar
A15046.0E |
2500 g |
499.49 €
2500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Crosslinking agent for modification of proteins via amidationMalononitrile is used as a precursor in the preparation of anti-cancer drug, triamterene, adenine, methotrexate, thiamine, acrylic fibers and dyes. It is used in Knoevenagel condensation to prepare 2-chlorobenzalmalononitrile by reacting with 2-chlorobenzaldehyde. It acts as a leaching agent for gold. It is used as a raw material for the Gewald reaction of ketone or aldehyde to produce 2-aminothiophene using elemental sulfur and a base. Further, it plays an important role in the preparation of [1,6]-naphthyridines, gamma-ketoamides, 1,4-dihydropyridine, chromeno[2,3-b]pyridine and dihydro-1,4-dithiepine frameworks.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsCrosslinking agent for modification of proteins via amidationMalononitrile is used as a precursor in the preparation of anti-cancer drug, triamterene, adenine, methotrexate, thiamine, acrylic fibers and dyes. It is used in Knoevenagel condensation to prepare 2-chlorobenzalmalononitrile by reacting with 2-chlorobenzaldehyde. It acts as a leaching agent for gold. It is used as a raw material for the Gewald reaction of ketone or aldehyde to produce 2-aminothiophene using elemental sulfur and a base. Further, it plays an important role in the preparation of [1,6]-naphthyridines, gamma-ketoamides, 1,4-dihydropyridine, chromeno[2,3-b]pyridine and dihydro-1,4-dithiepine frameworks.
Solubility
Soluble in water, acetone, acetic acid, chloroform, ethanol, ether and benzene.
Notes
Store in a cool place. Incompatible with strong oxidizing agents, strong reducing agents, strong acids and strong bases.
Chemical Identifiers
109-77-3 | |
MFCD00001883 | |
malononitrile, dicyanomethane, cyanoacetonitrile, malonic dinitrile, methylene cyanide, dicyanmethane, malonodinitrile, malonic acid dinitrile, methylenedinitrile, methane, dicyano | |
CHEBI:33186 |
C3H2N2 | |
CUONGYYJJVDODC-UHFFFAOYSA-N | |
8010 | |
propanedinitrile |
Specifications
109-77-3 | |
1.049 | |
112°C | |
1.415 | |
UN2647 | |
14,5711 | |
Soluble aq. soln.,acetone,acetic acid,chloroform,ethanol,ether and benzene. | |
N#CCC#N | |
66.06 | |
CHEBI:33186 | |
99% |
30°C to 34°C | |
220°C to 222°C | |
C3H2N2 | |
MFCD00001883 | |
773697 | |
malononitrile, dicyanomethane, cyanoacetonitrile, malonic dinitrile, methylene cyanide, dicyanmethane, malonodinitrile, malonic acid dinitrile, methylenedinitrile, methane, dicyano | |
CUONGYYJJVDODC-UHFFFAOYSA-N | |
propanedinitrile | |
8010 | |
66.06 | |
Malononitrile |
Safety and Handling
GHS H Statement
H301-H311-H331
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P311-P312-P330-P361-P363-P501c
H301+H311+H331
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: MALONONITRILE
missing translation for 'einecsNumber' : 203-703-2
missing translation for 'rtecsNumber' : OO3150000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
- Malononitrile
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 2
- Acute toxicity Category 3
- LONG-TERM AQUATIC HAZARD Chronic 1
- Serious eye damage/eye irritation Category 2
- Skin sensitiser Category 1
Hazard Statement
- H300-Fatal if swallowed.
- H311-Toxic in contact with skin.
- H317-May cause an allergic skin reaction.
- H319-Causes serious eye irritation.
- H331-Toxic if inhaled.
- H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P311-Call a POISON CENTER/doctor/ .
Supplemental information
- MIXTURE LIST-Contains : Propanedinitrile
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.