missing translation for 'onlineSavingsMsg'
Learn More

Mancozeb, Thermo Scientific™

Product Code. 17545555 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 17545555

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 8018-01-7
Molecular Formula C8H12MnN4S8Zn
Molecular Weight (g/mol) 541.01
MDL Number 00078616
InChI Key CHNQZRKUZPNOOH-UHFFFAOYSA-J
IUPAC Name manganese(2+) zinc(2+) bis(({2-[(sulfanidylmethanethioyl)amino]ethyl}carbamothioyl)sulfanide)
SMILES [Mn++].[Zn++].[S-]C(=S)NCCNC([S-])=S.[S-]C(=S)NCCNC([S-])=S

Specifications

Packaging Glass Bottle
Certificate of Analysis Available
Physical Form Powder
Color Yellow to Gray
Merck Index 13-5738
Quantity 1 g
Chemical Name or Material Mancozeb
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.