missing translation for 'onlineSavingsMsg'
Learn More

Manganese Chloride, 1.0M Solution (Clear, Lt.-Pink Solution/Molecular Biology), Fisher BioReagents

Product Code. 10306843
Click to view available options
Poly Bottle; 100mL
This item is not returnable. View return policy
This item is not returnable. View return policy
Manganese Chloride is used for dC and dG tailing of DNA.
TRUSTED_SUSTAINABILITY

Specifications

Color Red
CAS 7773-01-5
Molecular Formula Cl2Mn
MDL Number MFCD00011114
Concentration 0.99 to 1.01M
Merck Index 15, 5793
Synonym manganese chloride, manganese ii chloride, manganese 2+ ion dichloride, manganesechloride, acmc-20akkt, manganous chloride,anhydrous, ksc171o6h, manganous chloride, anhydr, manganese ii chloride anhydrous crystalline
InChI Key GLFNIEUTAYBVOC-UHFFFAOYSA-L
SMILES [Cl-].[Cl-].[Mn++]
IUPAC Name manganese(2+) dichloride
Molecular Weight (g/mol) 125.84
PubChem CID 10313134
ChEBI CHEBI:63041
Grade Molecular Biology
Identification Pass Test
Packaging Poly Bottle
pH 4
Quantity 100 mL
Physical Form Liquid
Chemical Name or Material Manganese Chloride, 1.0M Solution
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Manganese chloride 1.0M solution
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 1
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H373-May cause damage to organs.
  • H412-Harmful to aquatic life with long lasting effects.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P311-Call a POISON CENTER/doctor/ .
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.