missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Manganese granules, 0.8-12mm (0.03-0.47in), 99.98% (metals basis), Thermo Scientific Chemicals
106.53 € - 305.52 €
Chemical Identifiers
CAS | 7439-96-5 |
---|---|
Molecular Formula | Mn |
MDL Number | MFCD00011111 |
InChI Key | PWHULOQIROXLJO-UHFFFAOYSA-N |
Synonym | colloidal, mangan, manganese, elemental, cutaval, magnacat, metal alloy, tronamang, fume, mangan polish, compounds |
PubChem CID | 23930 |
ChEBI | CHEBI:35154 |
IUPAC Name | manganese |
SMILES | [Mn] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11363258
|
Thermo Scientific Alfa Aesar
036215.14 |
25 g |
106.53 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11353258
|
Thermo Scientific Alfa Aesar
036215.22 |
100 g |
305.52 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
For vacuum deposition
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7439-96-5 | |
MFCD00011111 | |
colloidal, mangan, manganese, elemental, cutaval, magnacat, metal alloy, tronamang, fume, mangan polish, compounds | |
CHEBI:35154 | |
[Mn] |
Mn | |
PWHULOQIROXLJO-UHFFFAOYSA-N | |
23930 | |
manganese |
Specifications
7439-96-5 | |
Mn | |
colloidal, mangan, manganese, elemental, cutaval, magnacat, metal alloy, tronamang, fume, mangan polish, compounds | |
[Mn] | |
54.94 | |
CHEBI:35154 | |
Manganese |
99.98% (metals basis) | |
MFCD00011111 | |
PWHULOQIROXLJO-UHFFFAOYSA-N | |
manganese | |
23930 | |
Granular |
Safety and Handling
missing translation for 'einecsNumber' : 231-105-1
missing translation for 'rtecsNumber' : OO9275000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only