missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Manganese(II) acetate, 98+%, anhydrous, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Color | Beige-Gray to Brown |
Physical Form | Powder |
Quantity | 100 g |
Assay Percent Range | 98% min. (Complexometry) |
Linear Formula | Mn(OC(O)CH3)2 |
Merck Index | 15, 5789 |
Solubility Information | Solubility in water: very soluble |
Formula Weight | 173.03 |
Percent Purity | ≥98% |
Infrared Spectrum | Authentic |
Show More |
Safety and Handling
Product Identifier
- Manganese(II) acetate
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'