missing translation for 'onlineSavingsMsg'
Learn More

Manganese(II) acetate, 98+%, anhydrous, Thermo Scientific Chemicals

Product Code. 10782264
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 638-38-0
Molecular Formula C4H6MnO4
Molecular Weight (g/mol) 173.03
MDL Number MFCD00013039
InChI Key UOGMEBQRZBEZQT-UHFFFAOYSA-L
Synonym manganese ii acetate, manganese acetate, manganous acetate, diacetylmanganese, manganese diacetate, manganese di acetate, manganese 2+ acetate, octan manganaty czech, acetic acid, manganese 2+ salt, unii-0v6e9q2i0y
PubChem CID 12525
IUPAC Name manganese(2+);diacetate
SMILES [Mn++].CC([O-])=O.CC([O-])=O

Specifications

Color Beige-Gray to Brown
Physical Form Powder
Quantity 25 g
Assay Percent Range 98% min. (Complexometry)
Linear Formula Mn(OC(O)CH3)2
Merck Index 15, 5789
Solubility Information Solubility in water: very soluble
Formula Weight 173.03
Percent Purity ≥98%
Infrared Spectrum Authentic
Packaging Glass bottle
Chemical Name or Material Manganese(II) acetate
Show More Show Less
Product Identifier
  • Manganese(II) acetate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'