missing translation for 'onlineSavingsMsg'
Learn More

Manganese(II) bromide, anhydrous, 95%, Thermo Scientific Chemicals

Product Code. 11423887
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13446-03-2
Molecular Formula MnBr2
MDL Number MFCD00011113

Specifications

Melting Point 698°C
Quantity 25 g
Merck Index 14,5725
Formula Weight 214.76
Percent Purity 95%
Odor Odorless
Sensitivity Hygroscopic
Density 4.38
Chemical Name or Material Manganese(II) bromide, anhydrous

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'