Learn More
Manganese(II) carbonate, 99.9% (metals basis excluding Na), Thermo Scientific Chemicals
57.10 € - 152.00 €
Chemical Identifiers
CAS | 598-62-9 |
---|---|
Molecular Formula | CMnO3 |
Molecular Weight (g/mol) | 114.946 |
MDL Number | MFCD00011116 |
InChI Key | XMWCXZJXESXBBY-UHFFFAOYSA-L |
Synonym | manganese carbonate, manganese ii carbonate, manganous carbonate, manganese 2+ carbonate, natural rhodochrosite, manganese carbonate 1:1, rhodochrosite, manganese carbonate mnco3, unii-9zv57512zm, ccris 3660 |
PubChem CID | 11726 |
IUPAC Name | manganese(2+);carbonate |
SMILES | C(=O)([O-])[O-].[Mn+2] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11317907
|
Thermo Scientific Alfa Aesar
014344.30 |
250 g |
57.10 €
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11307907
|
Thermo Scientific Alfa Aesar
014344.A1 |
1 kg |
152.00 €
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Manganese(II) carbonate is used as an additive to plant fertilizers, glaze colorant in ceramics, pigment and drier for varnishes. It is also used as an ingredient in welding rod, animal feed additive as well as to prepare other manganese salts and pharmaceuticals. It is also used in medicine as a hematinic and in health foods, in flux and in concrete stains.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Identifiants chimiques
598-62-9 | |
114.946 | |
XMWCXZJXESXBBY-UHFFFAOYSA-L | |
11726 | |
C(=O)([O-])[O-].[Mn+2] |
CMnO3 | |
MFCD00011116 | |
manganese carbonate, manganese ii carbonate, manganous carbonate, manganese 2+ carbonate, natural rhodochrosite, manganese carbonate 1:1, rhodochrosite, manganese carbonate mnco3, unii-9zv57512zm, ccris 3660 | |
manganese(2+);carbonate |
Spécification
350°C (decomposition) | |
Brown to Violet | |
99.9% (metals basis excluding Na) | |
MFCD00011116 | |
Soluble in dilute inorganic acids and carbon dioxide. Insoluble in water,alcohol,ammonia and common organic acids. | |
C(=O)([O-])[O-].[Mn+2] | |
114.946 | |
114.95 | |
Light brown to violet powder | |
Manganese(II) carbonate |
598-62-9 | |
Powder | |
CMnO3 | |
manganese carbonate, manganese ii carbonate, manganous carbonate, manganese 2+ carbonate, natural rhodochrosite, manganese carbonate 1:1, rhodochrosite, manganese carbonate mnco3, unii-9zv57512zm, ccris 3660 | |
XMWCXZJXESXBBY-UHFFFAOYSA-L | |
manganese(2+);carbonate | |
11726 | |
Odorless | |
3.125 g/cm3 |
Sécurité et traitement
missing translation for 'einecsNumber' : 209-942-9
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only