Learn More
Manganese(II) carbonate, Puratronic™, 99.985% (metals basis), Thermo Scientific Chemicals
Description
Manganese(II) carbonate is used in the production of manganese dioxide for dry-cell batteries and for ferrites, as an additive to plant fertilizers to cure manganese deficient crops. It is also used in health foods, in ceramics as a glaze colorant and flux, and in concrete stains, and as a hematinic.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Spécification
Spécification
Melting Point | 350°C (decomposition) |
Color | Brown to Violet |
Physical Form | Powder |
Quantity | 5 g |
Assay Percent Range | 99.99% (metals basis) |
Solubility Information | Soluble in water(0.065g/L),dilute inorganic acids. Insoluble alcohol. |
Formula Weight | 114.95 |
Odor | Odorless |
Appearance | Light brown to violet powder |
Density | 3.125 |
Afficher plus |
RUO – Research Use Only
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.