missing translation for 'onlineSavingsMsg'
Learn More

Manganese(II) carbonate, Puratronic™, 99.985% (metals basis), Thermo Scientific Chemicals

38.79 € - 514.56 €

Chemical Identifiers

CAS 598-62-9
Molecular Formula CMnO3
Molecular Weight (g/mol) 114.946
MDL Number MFCD00011116
InChI Key XMWCXZJXESXBBY-UHFFFAOYSA-L
Synonym manganese carbonate, manganese ii carbonate, manganous carbonate, manganese 2+ carbonate, natural rhodochrosite, manganese carbonate 1:1, rhodochrosite, manganese carbonate mnco3, unii-9zv57512zm, ccris 3660
PubChem CID 11726
IUPAC Name manganese(2+);carbonate
SMILES C(=O)([O-])[O-].[Mn+2]
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11345057
View Documents
Thermo Scientific Alfa Aesar
040756.06
5 g
38.79 €
5g
Please to purchase this item. Need a web account? Register with us today!
11335057
View Documents
Thermo Scientific Alfa Aesar
040756.14
25 g
153.77 €
25g
Please to purchase this item. Need a web account? Register with us today!
11355057
View Documents
Thermo Scientific Alfa Aesar
040756.22
100 g
514.56 €
100g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Manganese(II) carbonate is used in the production of manganese dioxide for dry-cell batteries and for ferrites, as an additive to plant fertilizers to cure manganese deficient crops. It is also used in health foods, in ceramics as a glaze colorant and flux, and in concrete stains, and as a hematinic.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

598-62-9
114.946
XMWCXZJXESXBBY-UHFFFAOYSA-L
11726
C(=O)([O-])[O-].[Mn+2]
CMnO3
MFCD00011116
manganese carbonate, manganese ii carbonate, manganous carbonate, manganese 2+ carbonate, natural rhodochrosite, manganese carbonate 1:1, rhodochrosite, manganese carbonate mnco3, unii-9zv57512zm, ccris 3660
manganese(2+);carbonate

Specifications

350°C (decomposition)
Brown to Violet
99.99% (metals basis)
MFCD00011116
Soluble in water(0.065g/L),dilute inorganic acids. Insoluble alcohol.
C(=O)([O-])[O-].[Mn+2]
114.946
114.95
Light brown to violet powder
Manganese(II) carbonate
598-62-9
Powder
CMnO3
manganese carbonate, manganese ii carbonate, manganous carbonate, manganese 2+ carbonate, natural rhodochrosite, manganese carbonate 1:1, rhodochrosite, manganese carbonate mnco3, unii-9zv57512zm, ccris 3660
XMWCXZJXESXBBY-UHFFFAOYSA-L
manganese(2+);carbonate
11726
Odorless
3.125
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 209-942-9

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only