missing translation for 'onlineSavingsMsg'
Learn More
Manganese(II) chloride, 97%, Thermo Scientific Chemicals Product Code.: 11381938

Manganese(II) chloride, 97%, Thermo Scientific Chemicals

Product Code. 11381938
2 kg
Click to view available options
Quantity:
500 g
2 kg
10 kg
This item is not returnable. View return policy

Product Code. 11381938

Brand: Thermo Scientific Alfa Aesar 011868.A3

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Manganese(II) chloride, plays an important role in the production of dry cell batteries. It is also used as a starting material to the methylcyclopentadienyl manganese tricarbonyl, which has antiknock property. Further, it is used in the formation of an active Manganese(0) species useful for radical cyclization. It is also used in 31P-NMR to determine the size and lamellarity of phospholipidvesicles reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7773-01-5
Molecular Formula Cl2Mn
Molecular Weight (g/mol) 125.84
MDL Number MFCD00011114
InChI Key GLFNIEUTAYBVOC-UHFFFAOYSA-L
Synonym manganese chloride, manganese ii chloride, manganese 2+ ion dichloride, manganesechloride, acmc-20akkt, manganous chloride,anhydrous, ksc171o6h, manganous chloride, anhydr, manganese ii chloride anhydrous crystallineShow MoreShow Less
PubChem CID 10313134
ChEBI CHEBI:63041
SMILES [Cl-].[Cl-].[Mn++]

Specifications

Boiling Point 1190°C
Melting Point 650°C
Color Pink
Physical Form Crystalline Flakes
Quantity 2 kg
Assay Percent Range 97%
Solubility Information Soluble in water,pyridine and alcohol. Insoluble in ether.
IUPAC Name manganese(2+);dichloride
Formula Weight 125.84
Odor Odorless
Sensitivity Hygroscopic
Density 2.977
Chemical Name or Material Manganese(II) chloride
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberM20L007 Certificate TypeCertificate of Analysis Date22/01/2025 Catalog Number011868.A3,011868.36,011868.A9
Lot NumberM20L006 Certificate TypeCertificate of Analysis Date22/01/2025 Catalog Number011868.A3,011868.36,011868.A9
Lot NumberZ14D025 Certificate TypeCertificate of Analysis Date31/10/2022 Catalog Number011868.A3,011868.36,011868.A9
Lot NumberW06C015 Certificate TypeCertificate of Analysis Date31/10/2022 Catalog Number011868.A3,011868.36,011868.A9
Lot NumberI12Y016 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog Number011868.A3,011868.36,011868.A9

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Manganese(II) chloride, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.