missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Manganese(II) chloride, 99+%, anhydrous, -80 mesh, Thermo Scientific Chemicals
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Boiling Point | 1190°C |
Melting Point | 650°C |
Color | Pink |
Physical Form | Powder |
Quantity | 10 g |
Assay Percent Range | 99% min. (Complexometry) |
Linear Formula | MnCl2 |
Fieser | 07,222 |
Merck Index | 15, 5793 |
Solubility Information | Solubility in water: 723g/L (25°C). Other solubilities: soluble in alcohol,insoluble in ether |
Show More |
Safety and Handling
Product Identifier
- Manganese(II) chloride
- Danger
- Acute toxicity Category 3
- LONG-TERM AQUATIC HAZARD Chronic 2
- Serious eye damage/eye irritation Category 1
- Specific target organ toxicity after repeated exposure Category 2
- H301-Toxic if swallowed.
- H318-Causes serious eye damage.
- H373-May cause damage to organs.
- H411-Toxic to aquatic life with long lasting effects.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- MIXTURE LIST-Contains : Manganese(II) chloride
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction