Learn More
Manganese(II) chloride tetrahydrate, ACS reagent, Thermo Scientific Chemicals
Chemical Identifiers
CAS | 13446-34-9 |
---|---|
Molecular Formula | Cl2H8MnO4 |
Molecular Weight (g/mol) | 197.90 |
MDL Number | MFCD00149792 |
InChI Key | CNFDGXZLMLFIJV-UHFFFAOYSA-L |
IUPAC Name | manganese(2+) tetrahydrate dichloride |
SMILES | O.O.O.O.[Cl-].[Cl-].[Mn++] |
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
13446-34-9 | |
197.90 | |
CNFDGXZLMLFIJV-UHFFFAOYSA-L | |
O.O.O.O.[Cl-].[Cl-].[Mn++] |
Cl2H8MnO4 | |
MFCD00149792 | |
manganese(2+) tetrahydrate dichloride |
Specifications
58.0°C | |
Cl2H8MnO4 | |
MFCD00149792 | |
15,5793 | |
CNFDGXZLMLFIJV-UHFFFAOYSA-L | |
manganese(2+) tetrahydrate dichloride | |
197.91 | |
Manganese(II) chloride tetrahydrate |
13446-34-9 | |
MnCl2·4H2O | |
07,222; 09,288 | |
Solubility in water: 1980g/L (20°C). Other solubilities: soluble in alcohol, insoluble in ether | |
O.O.O.O.[Cl-].[Cl-].[Mn++] | |
197.90 | |
ACS Reagent |
Safety and Handling
GHS H Statement
May cause damage to organs through prolonged or repeated exposure if inhaled.
Harmful if swallowed.
Toxic to aquatic life with long lasting effects.
Causes serious eye damage.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre
GHS Signal Word: Danger
- Manganese(II) chloride tetrahydrate
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 2
- Serious eye damage/eye irritation Category 1
- Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
- H318-Causes serious eye damage.
- H373-May cause damage to organs.
- H411-Toxic to aquatic life with long lasting effects.
- H302-Harmful if swallowed.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only