missing translation for 'onlineSavingsMsg'
Learn More

Manganese(II) chloride, ultra dry, 99.99% (metals basis), Thermo Scientific Chemicals

103.52 € - 954.75 €

Chemical Identifiers

CAS 7773-01-5
Molecular Formula Cl2Mn
Molecular Weight (g/mol) 125.84
MDL Number MFCD00011114
InChI Key GLFNIEUTAYBVOC-UHFFFAOYSA-L
Synonym manganese chloride, manganese ii chloride, manganese 2+ ion dichloride, manganesechloride, acmc-20akkt, manganous chloride,anhydrous, ksc171o6h, manganous chloride, anhydr, manganese ii chloride anhydrous crystalline
PubChem CID 10313134
ChEBI CHEBI:63041
SMILES [Cl-].[Cl-].[Mn++]
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11322418
View Documents
Thermo Scientific Alfa Aesar
014464.06
5 g
103.52 €
5g
Please to purchase this item. Need a web account? Register with us today!
11342418
View Documents
Thermo Scientific Alfa Aesar
014464.14
25 g
348.74 €
25g
Please to purchase this item. Need a web account? Register with us today!
11332418
View Documents
Thermo Scientific Alfa Aesar
014464.22
100 g
954.75 €
100g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Manganese(II) chloride is used in formation of an active Mn(0) species useful for radical cyclization reactions. It is used as catalyst, raw material for paint drier and antiknock agent for gasoline. It iis also used in the production of pigments, dyes, car batteries, chemical reagents and trace elements.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
Specifications

Chemical Identifiers

7773-01-5
125.84
GLFNIEUTAYBVOC-UHFFFAOYSA-L
10313134
[Cl-].[Cl-].[Mn++]
Cl2Mn
MFCD00011114
manganese chloride, manganese ii chloride, manganese 2+ ion dichloride, manganesechloride, acmc-20akkt, manganous chloride,anhydrous, ksc171o6h, manganous chloride, anhydr, manganese ii chloride anhydrous crystalline
CHEBI:63041

Specifications

1190°C
7773-01-5
Beads
MnCl2
UN3288
manganese chloride, manganese ii chloride, manganese 2+ ion dichloride, manganesechloride, acmc-20akkt, manganous chloride,anhydrous, ksc171o6h, manganous chloride, anhydr, manganese ii chloride anhydrous crystalline
GLFNIEUTAYBVOC-UHFFFAOYSA-L
manganese(2+);dichloride
10313134
125.84
Odorless
Hygroscopic
(metals basis)
650°C
Pink
Cl2Mn
MFCD00011114
14,5728
Soluble in water(723g/L at 25 C)
[Cl-].[Cl-].[Mn++]
125.84
CHEBI:63041
99.99%
Ampule
2.977 g/mL
Manganese(II) chloride, Ultra dry
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301
Toxic if swallowed.

P262-P264-P270-P280i-P301+P312-P305+P351+P338-P310-P314-P330-P501c

H302-H318-H373

missing translation for 'dotInformation' : Hazard Class: 9; Packaging Group: III

missing translation for 'einecsNumber' : 231-869-6

missing translation for 'rtecsNumber' : OO9625000

missing translation for 'tsca' : Yes

missing translation for 'storageNote' :

  • Ampouled under argon

Recommended Storage : Ambient temperatures; Store under Argon

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.