missing translation for 'onlineSavingsMsg'
Learn More

Manganese(II) methoxide, Thermo Scientific Chemicals

Product Code. 11328067
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Manganese(II) methoxide is used as a biochemical for proteomics research and it is also used in convenient for carboxylating active methylene compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7245-20-7
Molecular Formula C2H6MnO2
Molecular Weight (g/mol) 117.006
MDL Number MFCD00061476
InChI Key UQIRCVPINUNHQY-UHFFFAOYSA-N
Synonym manganese ii methoxide, acmc-20akkm, dimethoxymanganese ii, manganese ii bis methoxide, manganese 2+ bis methoxide, manganese 2+ ion bis methoxide
PubChem CID 10219413
IUPAC Name manganese(2+);methanolate
SMILES C[O-].C[O-].[Mn+2]

Specifications

Linear Formula Mn(OCH3)2
Quantity 5 g
UN Number UN3181
Sensitivity Moisture sensitive
Solubility Information Soluble in methanol
Formula Weight 117.01
Physical Form Powder
Chemical Name or Material Manganese(II) methoxide

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'