missing translation for 'onlineSavingsMsg'
Learn More

Manganese(II) nitrate hydrate, Puratronic™, 99.995% (metals basis), Thermo Scientific Chemicals

Product Code. 11356917
Click to view available options
Quantity:
2 g
10 g
50 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Raw material for electrochemical synthesis of a LaMnO3 thin film coating on stainless steel substrates. Also useful precursors to the oxides of manganese.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15710-66-4
Molecular Formula MnN2O6
Molecular Weight (g/mol) 178.95
MDL Number MFCD00149788
InChI Key MIVBAHRSNUNMPP-UHFFFAOYSA-N
Synonym manganese ii nitrate hydrate, acmc-1brrd, manganese nitrate hydrate, mn.2no3.h2o, manganese 2+ ion hydrate dinitrate, manganese 2+ hydrate dinitrate, manganese ii nitrate hydrate, puratronic, manganese 2+ nitrate-water 1/2/1
PubChem CID 16211480
SMILES [Mn++].[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

Melting Point ∽26°C
Color Pink
Physical Form Crystalline Aggregates
Quantity 2 g
Assay Percent Range 99.999% (Metal basis)
Solubility Information Soluble in water.
IUPAC Name manganese(2+);dinitrate;hydrate
Formula Weight 178.95 (Anhydrous)
Odor Odorless
Sensitivity Hygroscopic
Appearance Pink Crystalline Aggregates
Chemical Name or Material Manganese(II) nitrate hydrate
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.