missing translation for 'onlineSavingsMsg'
Learn More

Manganese(II) nitrate hydrate, Puratronic™, 99.995% (metals basis), Thermo Scientific Chemicals

Product Code. 11376917
Click to view available options
Quantity:
2 g
10 g
50 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Raw material for electrochemical synthesis of a LaMnO3 thin film coating on stainless steel substrates. Also useful precursors to the oxides of manganese.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15710-66-4
Molecular Formula MnN2O6
Molecular Weight (g/mol) 178.95
MDL Number MFCD00149788
InChI Key MIVBAHRSNUNMPP-UHFFFAOYSA-N
Synonym manganese ii nitrate hydrate, acmc-1brrd, manganese nitrate hydrate, mn.2no3.h2o, manganese 2+ ion hydrate dinitrate, manganese 2+ hydrate dinitrate, manganese ii nitrate hydrate, puratronic, manganese 2+ nitrate-water 1/2/1
PubChem CID 16211480
SMILES [Mn++].[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

Melting Point ∽26°C
Color Pink
Physical Form Crystalline Aggregates
Quantity 50 g
Assay Percent Range 99.999% (Metal basis)
Solubility Information Soluble in water.
IUPAC Name manganese(2+) dinitrate
Formula Weight 178.95 (anhydrous)
Odor Odorless
Sensitivity Hygroscopic
Appearance Pink Crystalline Aggregates
Chemical Name or Material Manganese(II) nitrate hydrate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'