missing translation for 'onlineSavingsMsg'
Learn More

Manganese(II) oxide, Mn 76.0-78.0%, typically 99%, Thermo Scientific Chemicals

Product Code. 11311948
missing translation for 'orderingAttributeHoverText'
Quantity:
500 g
2 kg
5 x 2 kg

The main use of manganese dioxide is in dry-cell batteries, such as the alkaline battery and the zinc-carbon battery. It is useful as a pigment, and as a precursor to other manganese compounds. It is used as a reagent in organic synthesis, for instance, for the oxidation of allylic alcohols and arylmethyl alcohols. It is also employed in various industrial purposes such as in adhesives and sealant chemicals, lubricants and lubricant additives, fertilizers, oxidizing/reducing agents, plating agents and surface treating agents, processing aids, air care products, paints, and coatings.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1344-43-0
Molecular Formula MnO
Molecular Weight (g/mol) 70.94
MDL Number MFCD00016226
InChI Key VASIZKWUTCETSD-UHFFFAOYSA-N
Synonym manganese ii oxide, manganosite, manganese monoxide, manganese green, cassel green, manganese oxide, rosensthiel green, nu-manese, manganese monooxide, manganese protoxide
PubChem CID 14940
IUPAC Name oxomanganese
SMILES O=[Mn]

Specifications

Color Green
Physical Form Powder
Quantity 5 x 2 kg
Assay Percent Range Mn 76.0-78.0%, typically 99%
Solubility Information Insoluble in water. Soluble in hydrochloric acid.
Formula Weight 70.94
Odor Odorless
Appearance Green powder
Density 5
Chemical Name or Material Manganese(II) oxide, Mn 76.0-78.0%

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'