missing translation for 'onlineSavingsMsg'
Learn More

Manganese(II) oxide, Mn 76.0-78.0%, typically 99%

Product Code. p-3257043
Click to view available options
Quantity:
500 g
2 kg
5 x 2 kg
Unit Size:
10kg
2kg
500g
This item is not returnable. View return policy

Product Code. 11331948

Brand: Thermo Scientific Alfa Aesar 011870.36

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1344-43-0 | MnO | 70.94 g/mol

The main use of manganese dioxide is in dry-cell batteries, such as the alkaline battery and the zinc-carbon battery. It is useful as a pigment, and as a precursor to other manganese compounds. It is used as a reagent in organic synthesis, for instance, for the oxidation of allylic alcohols and arylmethyl alcohols. It is also employed in various industrial purposes such as in adhesives and sealant chemicals, lubricants and lubricant additives, fertilizers, oxidizing/reducing agents, plating agents and surface treating agents, processing aids, air care products, paints, and coatings.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1344-43-0
Molecular Formula MnO
Molecular Weight (g/mol) 70.94
MDL Number MFCD00016226
InChI Key VASIZKWUTCETSD-UHFFFAOYSA-N
Synonym manganese ii oxide, manganosite, manganese monoxide, manganese green, cassel green, manganese oxide, rosensthiel green, nu-manese, manganese monooxide, manganese protoxide
PubChem CID 14940
IUPAC Name oxomanganese
SMILES O=[Mn]

Specifications

Color Green
Physical Form Powder
Quantity 500 g
Assay Percent Range Mn 76.0-78.0%, typically 99%
Solubility Information Insoluble in water. Soluble in hydrochloric acid.
Formula Weight 70.94
Odor Odorless
Appearance Green powder
Density 5 g/cm3 at 20°C
Chemical Name or Material Manganese(II) oxide, Mn 76.0-78.0%

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.