Learn More
Manganese(II) sulfate hydrate, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals
53.15 € - 165.83 €
Chemical Identifiers
CAS | 15244-36-7 |
---|---|
Molecular Formula | H2MnO5S |
Molecular Weight (g/mol) | 169.01 |
MDL Number | MFCD00149159 |
InChI Key | ISPYRSDWRDQNSW-UHFFFAOYSA-L |
Synonym | manganese sulfate monohydrate, manganese ii sulfate monohydrate, manganese sulfate hydrate, manganese ii sulfate hydrate, mnso4.h2o, manganous sulfate monohydrate, unii-w00lys4t26, manganese 2+ sulfate monohydrate, manganese sulfate usp, manganese 2+ sulfate hydrate |
PubChem CID | 177577 |
ChEBI | CHEBI:86364 |
SMILES | O.[Mn++].[O-]S([O-])(=O)=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11396917
|
Thermo Scientific Alfa Aesar
010807.06 |
5 g | N/A | ||||||
11386917
|
Thermo Scientific Alfa Aesar
010807.14 |
25 g |
165.83 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Used in feed additive, nutrient, firming agent and flavor enhancer. It is also used as a fermentation aid in the processing of beer and malt beverages.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
15244-36-7 | |
169.01 | |
ISPYRSDWRDQNSW-UHFFFAOYSA-L | |
177577 | |
O.[Mn++].[O-]S([O-])(=O)=O |
H2MnO5S | |
MFCD00149159 | |
manganese sulfate monohydrate, manganese ii sulfate monohydrate, manganese sulfate hydrate, manganese ii sulfate hydrate, mnso4.h2o, manganous sulfate monohydrate, unii-w00lys4t26, manganese 2+ sulfate monohydrate, manganese sulfate usp, manganese 2+ sulfate hydrate | |
CHEBI:86364 |
Specifications
15244-36-7 | |
Crystalline Powder | |
H2MnO5S | |
manganese sulfate monohydrate, manganese ii sulfate monohydrate, manganese sulfate hydrate, manganese ii sulfate hydrate, mnso4.h2o, manganous sulfate monohydrate, unii-w00lys4t26, manganese 2+ sulfate monohydrate, manganese sulfate usp, manganese 2+ sulfate hydrate | |
O.[Mn++].[O-]S([O-])(=O)=O | |
169.01 | |
CHEBI:86364 | |
Odorless | |
Manganese(II) sulfate hydrate, Loses all water at 400-450° |
Pink | |
99.999% (Metal basis) | |
MFCD00149159 | |
ISPYRSDWRDQNSW-UHFFFAOYSA-L | |
manganese(2+);sulfate;hydrate | |
177577 | |
151.00 (Anhydrous) | |
Pink Crystalline Powder |
Safety and Handling
H373-H411-H401: May cause damage to organs through prolonged or repeated exposure. Toxic to aquatic life with long lasting effects. Toxic to aquatic life.
missing translation for 'dotInformation' : DOT Class: 9, Packaging Group: III
missing translation for 'einecsNumber' : 232-089-9
RUO – Research Use Only