missing translation for 'onlineSavingsMsg'
Learn More

Manganese(II) sulfate monohydrate, 97%, Thermo Scientific Chemicals

Product Code. 11439403
Click to view available options
Quantity:
500 g
2500 g

Manganese(II) sulfate is used as a colorant in dyes. It is also used as feed additives, catalyst for the synthesis of chlorophyll, make plants grow well and increase production. It is used for making dryer of ink and paint. It is used in paper making , porcelain/ceramic, medicine and other industries.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Manganese(II) sulfate is used as a colorant in dyes. It is also used as feed additives, catalyst for the synthesis of chlorophyll, make plants grow well and increase production. It is used for making dryer of ink and paint. It is used in paper making , porcelain/ceramic, medicine and other industries.

Solubility
Soluble in water. 5-10 g/100 mL at 21°C

Notes
Loses all water at 400-450°
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10034-96-5
Molecular Formula H2MnO5S
Molecular Weight (g/mol) 169.01
MDL Number MFCD00149159
InChI Key ISPYRSDWRDQNSW-UHFFFAOYSA-L
Synonym manganese sulfate monohydrate, manganese ii sulfate monohydrate, manganese sulfate hydrate, manganese ii sulfate hydrate, mnso4.h2o, manganous sulfate monohydrate, unii-w00lys4t26, manganese 2+ sulfate monohydrate, manganese sulfate usp, manganese 2+ sulfate hydrate
PubChem CID 177577
ChEBI CHEBI:86364
SMILES O.[Mn++].[O-]S([O-])(=O)=O

Specifications

pH 3 to 3.5
Quantity 500 g
UN Number UN3077
Merck Index 14,5739
Solubility Information Soluble in water. 5-10g/100 mL at 21°C
IUPAC Name manganese(2+);sulfate;hydrate
Formula Weight 169.02 (151.00 Anhydrous)
Percent Purity 97%
Odor Odorless
Sensitivity Hygroscopic
Density 2.95
Chemical Name or Material Manganese (II) sulfate monohydrate
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Manganese(II) sulfate monohydrate
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Serious eye damage/eye irritation Category 1
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H318-Causes serious eye damage.
  • H373-May cause damage to organs.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : Manganese sulfate monohydrate, Manganese(II) sulfate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'