missing translation for 'onlineSavingsMsg'
Learn More

Manganese(II) sulfate monohydrate, ACS, 98.0-101.0%, Thermo Scientific Chemicals

51.46 € - 137.69 €

Chemical Identifiers

CAS 10034-96-5
Molecular Formula H2MnO5S
Molecular Weight (g/mol) 169.01
MDL Number MFCD00149159
InChI Key ISPYRSDWRDQNSW-UHFFFAOYSA-L
Synonym manganese sulfate monohydrate, manganese ii sulfate monohydrate, manganese sulfate hydrate, manganese ii sulfate hydrate, mnso4.h2o, manganous sulfate monohydrate, unii-w00lys4t26, manganese 2+ sulfate monohydrate, manganese sulfate usp, manganese 2+ sulfate hydrate
PubChem CID 177577
ChEBI CHEBI:86364
IUPAC Name manganese(2+);sulfate;hydrate
SMILES O.[Mn++].[O-]S([O-])(=O)=O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11357668
View Documents Promotion Details
Thermo Scientific Alfa Aesar
033341.36
500 g
51.46 €
500g
Please to purchase this item. Need a web account? Register with us today!
11367668
View Documents Promotion Details
Thermo Scientific Alfa Aesar
033341.A3
2 kg
137.69 €
2kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Manganese(II) sulfate monohydrate is used as a colorant in dyes, fertilizers, animal feeds and red glazes on porcelain. Further, it is used in paints, ceramics, nutrient and dietary supplement. It is involved in the preparation of manganese dioxide. In addition, it serves as a precursor to manganese metal and other manganese compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

10034-96-5
169.01
ISPYRSDWRDQNSW-UHFFFAOYSA-L
177577
manganese(2+);sulfate;hydrate
H2MnO5S
MFCD00149159
manganese sulfate monohydrate, manganese ii sulfate monohydrate, manganese sulfate hydrate, manganese ii sulfate hydrate, mnso4.h2o, manganous sulfate monohydrate, unii-w00lys4t26, manganese 2+ sulfate monohydrate, manganese sulfate usp, manganese 2+ sulfate hydrate
CHEBI:86364
O.[Mn++].[O-]S([O-])(=O)=O

Specifications

10034-96-5
Powder
H2MnO5S
manganese sulfate monohydrate, manganese ii sulfate monohydrate, manganese sulfate hydrate, manganese ii sulfate hydrate, mnso4.h2o, manganous sulfate monohydrate, unii-w00lys4t26, manganese 2+ sulfate monohydrate, manganese sulfate usp, manganese 2+ sulfate hydrate
ISPYRSDWRDQNSW-UHFFFAOYSA-L
manganese(2+);sulfate;hydrate
177577
169.02 (151.00 Anhydrous)
Pink powder
Manganese(II) sulfate monohydrate
Pink
98.0-101.0%
MFCD00149159
Soluble in water and alcohol. Insoluble in ether.
O.[Mn++].[O-]S([O-])(=O)=O
169.01
CHEBI:86364
ACS
2.95
Videos
Safety and Handling

Safety and Handling

P260-P280-P305+P351+P338-P310-P314-P501c

H318-H373

missing translation for 'dotInformation' : DOT Class: 9; Packing Group: III

missing translation for 'einecsNumber' : 232-089-9

missing translation for 'rtecsNumber' : OP0893500

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

Product Identifier
  • Manganese(II) sulfate monohydrate

Signal Word
  • Danger

Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Serious eye damage/eye irritation Category 1
  • Specific target organ toxicity after repeated exposure Category 2

Hazard Statement
  • H318-Causes serious eye damage.
  • H373-May cause damage to organs.
  • H411-Toxic to aquatic life with long lasting effects.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

Supplemental information
  • MIXTURE LIST-Contains : Manganese sulfate monohydrate

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only