missing translation for 'onlineSavingsMsg'
Learn More

Manganese(II) sulfate monohydrate, Puriss. p.a., ACS Reagent, Reag. Ph. Eur., 99.0-101.0%, Honeywell Fluka™

Product Code. 15603400
Click to view available options
Quantity:
500 g
1 kg
50 kg
6 x 1 kg
This item is not returnable. View return policy
This item is not returnable. View return policy

Puriss. p.a., ACS Reagent, Reag. Ph. Eur., 99.0-101.0%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10034-96-5
Molecular Formula H2MnO5S
Molecular Weight (g/mol) 169.01
MDL Number MFCD00149159
InChI Key ISPYRSDWRDQNSW-UHFFFAOYSA-L
Synonym manganese sulfate monohydrate, manganese ii sulfate monohydrate, manganese sulfate hydrate, manganese ii sulfate hydrate, mnso4.h2o, manganous sulfate monohydrate, unii-w00lys4t26, manganese 2+ sulfate monohydrate, manganese sulfate usp, manganese 2+ sulfate hydrate
PubChem CID 177577
ChEBI CHEBI:86364
IUPAC Name manganese(2+);sulfate;hydrate
SMILES O.[Mn++].[O-]S([O-])(=O)=O

Specifications

Quantity 1 kg
Linear Formula MnSO4 · H2O
UN Number 3077
Formula Weight 169.02g/mol
Percent Purity 99 to 101%
Grade ACS/puriss. p.a.
Chemical Name or Material Manganese(Ii) Sulfate Monohydrate, Reag. Ph. Eur.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'