Learn More
Manganese(II) sulfide, 99.9% (metals basis), Thermo Scientific Chemicals
43.52 € - 646.22 €
Chemical Identifiers
CAS | 18820-29-6 |
---|---|
Molecular Formula | MnS |
Molecular Weight (g/mol) | 87.00 |
MDL Number | MFCD00014210 |
InChI Key | VCTOKJRTAUILIH-UHFFFAOYSA-N |
Synonym | manganese sulfide, manganese ii sulfide, manganous sulfide, manganese sulphide, manganese 2+ sulfide, 12687-82-0 cpd with unspecified mf, manganese ii sulfide, ∼100 mesh particle size |
PubChem CID | 87809 |
SMILES | [S--].[Mn++] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11357567
|
Thermo Scientific Alfa Aesar
012835.06 |
5 g |
43.52 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11347567
|
Thermo Scientific Alfa Aesar
012835.14 |
25 g |
183.92 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11337567
|
Thermo Scientific Alfa Aesar
012835.22 |
100 g |
646.22 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
it is used as an intermediate in chemical research& pharmaceutical research.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
18820-29-6 | |
87.00 | |
VCTOKJRTAUILIH-UHFFFAOYSA-N | |
87809 |
MnS | |
MFCD00014210 | |
manganese sulfide, manganese ii sulfide, manganous sulfide, manganese sulphide, manganese 2+ sulfide, 12687-82-0 cpd with unspecified mf, manganese ii sulfide, ∼100 mesh particle size | |
[S--].[Mn++] |
Specifications
1610°C (decomposition) | |
Green | |
99.9% (Metal basis) | |
MFCD00014210 | |
It is soluble in water. 0.47mg/100 mL (18°C). | |
[S--].[Mn++] | |
87.00 | |
87 | |
3.99 g/cm3 at 20°C |
18820-29-6 | |
Powder | |
MnS | |
manganese sulfide, manganese ii sulfide, manganous sulfide, manganese sulphide, manganese 2+ sulfide, 12687-82-0 cpd with unspecified mf, manganese ii sulfide, ∼100 mesh particle size | |
VCTOKJRTAUILIH-UHFFFAOYSA-N | |
sulfanylidenemanganese | |
87809 | |
Green powder | |
Manganese(II) sulfide |
Safety and Handling
H301: Toxic if swallowed.
missing translation for 'einecsNumber' : 242-599-3
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.