missing translation for 'onlineSavingsMsg'
Learn More

Manganese(II) tungsten oxide, 99.9% (metals basis), Thermo Scientific Chemicals

Product Code. 11308148
Click to view available options
Quantity:
50 g
250 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14177-46-9
MDL Number MFCD00014212
Synonym Manganese tungstate

Specifications

Physical Form Powder
Quantity 50 g
Solubility Information Insoluble in water.
Formula Weight 302.79
Percent Purity 99.9%
Odor Odorless
Density 7.23
Assay (metals basis)
Chemical Name or Material Manganese(II) tungsten oxide

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'