Learn More
Manganese(III) acetate dihydrate, 97%, Thermo Scientific Chemicals
Description
Manganese(III) acetate dihydrate, is used as a mild and selective oxidizing agent. It is also a reagent used for radical cyclizations and α-keto-acetoxylation. Mangese triacetate has been used as a single electron oxidant.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Color | Pink |
Physical Form | Solid |
Quantity | 50 g |
Linear Formula | Mn(OOCCH3)3·2H2O |
Beilstein | 3732626 |
Formula Weight | 268.10 (232.07 Anhydrous) |
Percent Purity | 97% |
Sensitivity | Hygroscopic |
Chemical Name or Material | Manganese(III) acetate dihydrate |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.