missing translation for 'onlineSavingsMsg'
Learn More

Manganese(III) acetate dihydrate, 98%

Product Code. p-44622 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 10420291

Brand: Thermo Scientific Acros 201740250

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 19513-05-4 | C6H9MnO6 | 232.07 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19513-05-4
Molecular Formula C6H9MnO6
Molecular Weight (g/mol) 232.07
MDL Number MFCD00150022
InChI Key AHSBSUVHXDIAEY-UHFFFAOYSA-K
IUPAC Name manganese(3+) triacetate
SMILES [Mn+3].CC([O-])=O.CC([O-])=O.CC([O-])=O

Specifications

Color Brown to Brown-Orange
Physical Form Powder
Quantity 25 g
Assay Percent Range 97.5% min. (Iodometry)
Linear Formula (CH3CO2)3Mn·2H2O
Fieser 02,263; 04,318; 06,355; 07,221; 14,197; 16,200; 17,175; 01,532; 02,233; 04,276
Solubility Information Solubility in water: very soluble
Formula Weight 268.09
Percent Purity 98%
Infrared Spectrum Authentic
Packaging Glass bottle
Chemical Name or Material Manganese(III) acetate dihydrate, 98%
Show More Show Less
compliance-icons
Product Identifier
  • Manganese(III) acetate dihydrate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H315-Causes skin irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P362-Take off contaminated clothing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.