missing translation for 'onlineSavingsMsg'
Learn More

Manganese(III) acetate hydrate, 96%, Thermo Scientific Chemicals

Product Code. 11387288
Click to view available options
Quantity:
10 g
50 g
This item is not returnable. View return policy
This item is not returnable. View return policy

It acts as a mild and selective oxidizing agent in organic synthesis and materials science. Manganese(III) Acetate Dihydrate is also a reagent used for radical cyclizations and α-keto-acetoxylation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19513-05-4
Molecular Formula C6H9MnO6
Molecular Weight (g/mol) 232.07
MDL Number MFCD00150022
InChI Key AHSBSUVHXDIAEY-UHFFFAOYSA-K
IUPAC Name manganese(3+) triacetate
SMILES [Mn+3].CC([O-])=O.CC([O-])=O.CC([O-])=O

Specifications

Color Pink
Physical Form Crystalline
Quantity 50 g
Linear Formula Mn(OOCCH3)3·xH2O
Solubility Information It is soluble in water.
Formula Weight 232.07 (Anhydrous)
Percent Purity 96%
Sensitivity Hygroscopic
Density 1.59 g/mL
Chemical Name or Material Manganese(III) acetate hydrate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'